
2,7-Dibromo-9,9-didecylfluorene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:175922-78-8
:
Formula:C₃₃H₄₈Br₂
MDL Number:MFCD00971288
:604.56 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:38.0 to 42.0 deg-C
Specification Test Results
Frequently Bought Together