Order Entry
Italy
ContactUsLinkComponent
2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene ≥98.0% (by HPLC, total nitrogen)
Irritant
2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
Irritant
2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    74.0 to 78.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together