
2,6-Dibromopyrazine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23229-25-6
:
Formula:C₄H₂Br₂N₂
MDL Number:MFCD08061601
Boiling Pt:65°C
:237.882 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:51.0 to 55.0 deg-C
Specification Test Results
Frequently Bought Together