
(4-Dimethylaminophenyl)di-tert-butylphosphine
Supplier: TCI
Certificates
About this item
CAS Number:932710-63-9
:
Formula:C₁₆H₂₈NP
MDL Number:MFCD09265102
:265.38 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 93.0 %
- Purity (Nonaqueous Titration):min. 93.0 %
- Purity (Iodometric Titration):min. 93.0 %
- Melting point:65.0 to 69.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together