Order Entry
Italy
ContactUsLinkComponent
4,7-Dibromo-2-(6-bromohexyl)benzotriazole ≥97.0% (by GC)
4,7-Dibromo-2-(6-bromohexyl)benzotriazole ≥97.0% (by GC)
Catalog # TCIAD4514-200MG
Supplier:  TCI
CAS Number:  
4,7-Dibromo-2-(6-bromohexyl)benzotriazole ≥97.0% (by GC)
Catalog # TCIAD4514-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :439.98 g/mol
CAS Number:890704-02-6
Formula:C₁₂H₁₄Br₃N₃
MDL Number:MFCD19440927