Order Entry
Italy
ContactUsLinkComponent
Diethyl-2,2':6',2''-terpyridine-4'-phosphonate ≥93.0% (by GC)
Irritant
Diethyl-2,2':6',2''-terpyridine-4'-phosphonate ≥93.0% (by GC)
Catalog # TCIAD4511-200MG
Supplier:  TCI
CAS Number:  
Irritant
Diethyl-2,2':6',2''-terpyridine-4'-phosphonate ≥93.0% (by GC)
Catalog # TCIAD4511-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 93.0 %
  • Melting point:
    147.0 to 151.0 deg-C

Specification Test Results

About this item

 :
 :369.36 g/mol
 :GHS07
CAS Number:161583-75-1
Formula:C₁₉H₂₀N₃O₃P
MDL Number:MFCD11042551