
3,6-Dimethylsalicylaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1666-04-2
:
Formula:C₉H₁₀O₂
MDL Number:MFCD04037328
:150.1700 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
Specification Test Results
Frequently Bought Together