(S)-(+)-DTBM-SEGPHOS® ≥99.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:210169-40-7
:
Formula:C₇₄H₁₀₀O₈P₂
MDL Number:MFCD09753003
:1179.530 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 99.0 area%
- Optical purity (LC):min. 99.0 ee%
Specification Test Results
Frequently Bought Together