Order Entry
Italy
ContactUsLinkComponent
 
(R)-(+)-DM-SEGPHOS® ≥95.0% (by HPLC)
 :  TCI
 
(R)-(+)-DM-SEGPHOS® ≥95.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Optical purity (LC):
    min. 99.0 ee%

 

Frequently Bought Together