(S)-(-)-DM-SEGPHOS® ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:210169-57-6
:
Formula:C₄₆H₄₄O₄P₂
MDL Number:MFCD09753008
:722.790 g/mol
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 95.0 area%
- Optical purity (LC):min. 99.0 ee%
Specification Test Results
Frequently Bought Together