Order Entry
Italy
ContactUsLinkComponent
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC, total nitrogen)
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    122.0 to 126.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together