
5,8-Dibromo-2,3-diphenylquinoxaline ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD4461-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:220.0 to 224.0 deg-C
Specification Test Results
About this item
:
:440.14 g/mol
:GHS07
CAS Number:94544-77-1
Formula:C₂₀H₁₂Br₂N₂
MDL Number:MFCD27966265