
3,3'-Diethyloxadicarbocyanine iodide (DODC) ≥98.0% (by total nitrogen basis)
Catalog # TCIAD4457-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Blue to Dark blue powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Absorbance (E1%1cm):min. 4500 (MeOH, 576.0 to 582.0 nm)
- Lambda max.:576.0 to 582.0 nm (MeOH)
Specification Test Results
About this item
:
:486.35 g/mol
EINECS:238-873-7
:GHS07
CAS Number:14806-50-9
Formula:C₂₃H₂₃IN₂O₂
MDL Number:MFCD00011953