
3,5-Dimethyl-4-nitropyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14531-55-6
:
Formula:C₅H₇N₃O₂
MDL Number:MFCD00052513
:141.13 g/mol
Boiling Pt:325°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:125.0 to 129.0 deg-C
Specification Test Results
Frequently Bought Together