Order Entry
Italy
ContactUsLinkComponent
Di-tert-butyl L-aspartate hydrochloride ≥98.0% (by HPLC, titration analysis)
Di-tert-butyl L-aspartate hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Di-tert-butyl L-aspartate hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +6.0 to +8.0 deg (C=1,MeOH)

Specification Test Results

Related Information

Frequently Bought Together