
2,2-Dimethylsuccinic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17347-61-4
:
Formula:C₆H₈O₃
MDL Number:MFCD00022608
:128.1259 g/mol
Boiling Pt:220°C
EINECS:241-371-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Methanolysis Method):min. 98.0 %
Specification Test Results
Frequently Bought Together