
3,6-Dibromo-2-picoline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39919-65-8
:
Formula:C₆H₅Br₂N
MDL Number:MFCD06254589
:250.92 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:36.0 to 40.0 deg-C
Specification Test Results
Frequently Bought Together