
2,3-Dichlorobenzonitrile ≥98.0% (by GC)
Catalog # TCIAD4398-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
About this item
:
:172.01 g/mol
UN:3439
ADR:6.1,III
:GHS06
CAS Number:6574-97-6
Formula:C₇H₃Cl₂N
MDL Number:MFCD00016372