Order Entry
Italy
ContactUsLinkComponent
 
 
Decahydro-1,4-naphthalenedicarboxylic acid (mixture of isomers) ≥96.0% (by GC, titration analysis)
  TCIAD4383-1G
 :  TCI
 :  
 
Decahydro-1,4-naphthalenedicarboxylic acid (mixture of isomers) ≥96.0% (by GC, titration analysis)
  TCIAD4383-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 96.0 % (total of isomers)
  • Purity (Neutralization titration):
    min. 96.0 %

 

 

 :
 :226.27 g/mol
 :GHS07
 :879360-14-2
 :C₁₂H₁₈O₄