Order Entry
Italy
ContactUsLinkComponent
5,5'-Dibromo-2,2'-bipyridyl ≥98.0% (by HPLC)
Irritant
5,5'-Dibromo-2,2'-bipyridyl ≥98.0% (by HPLC)
Catalog # TCIAD4358-1G
Supplier:  TCI
CAS Number:  
Irritant
5,5'-Dibromo-2,2'-bipyridyl ≥98.0% (by HPLC)
Catalog # TCIAD4358-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 96.0 %
  • Melting point:
    225.0 to 229.0 deg-C

Specification Test Results

About this item

 :
 :313.98 g/mol
 :GHS07
CAS Number:15862-18-7
Formula:C₁₀H₆Br₂N₂
MDL Number:MFCD06657883