Order Entry
Italy
ContactUsLinkComponent
1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol hydrochloride ≥98.0% (by HPLC, titration analysis)
1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4355-200MG
Supplier:  TCI
CAS Number:  
1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4355-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    117.0 to 121.0 deg-C

Specification Test Results

About this item

 :
 :365.90 g/mol
CAS Number:135261-74-4
Formula:C₂₀H₂₇NO₃
MDL Number:MFCD01697013