Order Entry
Italy
ContactUsLinkComponent
 
1-Decyl-3-methylimidazoliumbis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
1-Decyl-3-methylimidazoliumbis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %

 

Frequently Bought Together