
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD4324-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:135.13 g/mol
Melting Pt:345°C
:GHS07
Storage Temperature:Refrigerator
CAS Number:3680-71-5
:MFCD01686814
Formula:C₆H₅N₃O
MDL Number:MFCD08234923