

Dyclonine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:536-43-6
:
Formula:C₁₈H₂₈ClNO₂
MDL Number:MFCD00035386
Merck Index:14,03473
:325.877 g/mol
EINECS:208-633-6
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:171.0 to 175.0 deg-C
Specification Test Results
Frequently Bought Together