
2,3-Dimethylbenzonitrile ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5724-56-1
:
Formula:C₉H₉N
MDL Number:MFCD00019744
:131.18 g/mol
Boiling Pt:62°C
Melting Pt:16°C
EINECS:227-229-0
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together