
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:169051-76-7
:
Formula:C₁₀H₁₅F₆N₃O₄S₂
MDL Number:MFCD03788922
:419.37 g/mol
Melting Pt:8°C
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow clear liquid
- Purity (HPLC):min. 95.0 area%
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Frequently Bought Together