
2,2-Dibromomalonamide ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:73003-80-2
:
Formula:C₃H₄Br₂N₂O₂
MDL Number:MFCD00014803
:259.89 g/mol
EINECS:277-205-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:202.0 to 206.0 deg-C
Specification Test Results
Frequently Bought Together