
4,5-Diazafluoren-9-one ≥98.0% (by HPLC, titration analysis)
TCIAD4255-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:214.0 to 218.0 deg-C
:
:182.18 g/mol
:GHS07
:50890-67-0
:C₁₁H₆N₂O
:MFCD00046892