Order Entry
Italy
ContactUsLinkComponent
N,N'-Di-2-naphthyl-N,N'-diphenylbenzidine ≥99.0% (by HPLC, total nitrogen)
N,N'-Di-2-naphthyl-N,N'-diphenylbenzidine ≥99.0% (by HPLC, total nitrogen)
Catalog # TCIAD4253-1G
Supplier:  TCI
CAS Number:  
N,N'-Di-2-naphthyl-N,N'-diphenylbenzidine ≥99.0% (by HPLC, total nitrogen)
Catalog # TCIAD4253-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 99.0 %

Specification Test Results

About this item

 :
 :588.75 g/mol
Melting Pt:173°C
CAS Number:139255-17-7
Formula:C₄₄H₃₂N₂
MDL Number:MFCD03093247