Order Entry
Italy
ContactUsLinkComponent
2,6-Dichlorobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
Corrosive
2,6-Dichlorobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Corrosive
2,6-Dichlorobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Melting point:
    52.0 to 56.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together