
Di-tert-butyl-N,N-diisopropylphosphoramidite ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:137348-86-8
:
Formula:C₁₄H₃₂NO₂P
MDL Number:MFCD00153506
:277.39 g/mol
Boiling Pt:77°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together