Drospirenone ≥98.0% (by HPLC)
TCIAD4209-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:199.0 to 203.0 deg-C
- Specific rotation [a]20/D:-185.0 to -191.0 deg (C=1、MeOH)
:
:366.50 g/mol
:14,03454
:Refrigerator
:266-679-2
:67392-87-4
:C₂₄H₃₀O₃
:MFCD00867350