
1,5-Dichloro-2,4-dinitrobenzene ≥98.0% (by GC)
TCIAD4205-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:100.0 to 104.0 deg-C
:
:237 g/mol
:223-027-1
:3077
:9,III
:GHS09
:3698-83-7
:C₆H₂Cl₂N₂O₄
:MFCD00024186