Phenylboronic acid neopentyl glycol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5123-13-7
:
Formula:C₁₁H₁₅BO₂
MDL Number:MFCD05663851
:190.05 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together