1,3-Dibromo-7-tert-butylpyrene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1005771-04-9
:
Formula:C₂₀H₁₆Br₂
MDL Number:MFCD22581440
:416.16 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together