Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethylvaleronitrile
2,2-Dimethylvaleronitrile ≥98.0% (by GC)
2,2-Dimethylvaleronitrile ≥98.0% (by GC)
TCIAD4171-5G
:
TCI
:
2,2-Dimethylvaleronitrile ≥98.0% (by GC)
TCIAD4171-5G
:
TCI
:
Pk:
5 g
Seq:
3173
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 98.0 %
:
:
111.19 g/mol
:
150°C
:
1993
:
3,III
:
GHS02
:
GHS07
:
20654-47-1
:
C₇H₁₃N