Order Entry
Italy
ContactUsLinkComponent
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light gray to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Frequently Bought Together