2-(3,6-Dihexyloxyxanthen-9-yl)benzamide ≥97.0% (by HPLC, total nitrogen)
TCIAD4142-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:125.0 to 129.0 deg-C
:
:501.67 g/mol
:1820618-83-4
:C₃₂H₃₉NO₄