3,5-Di-tert-butylbenzaldehyde ≥97.0% (by GC)
: TCI
:17610-00-3
:
:C₁₅H₂₂O
:MFCD00026298
:132°C
:218.34 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:84.0 to 88.0 deg-C
Frequently Bought Together