Order Entry
Italy
ContactUsLinkComponent
 
 
Di-tert-butyl Iminodiacetate ≥97.0% (by GC)
 :  TCI
 
 
Di-tert-butyl Iminodiacetate ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    40.0 to 44.0 deg-C

 

Frequently Bought Together