1,3-Di-9-carbazolylbenzene ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Catalog # TCIAD4087-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:182.0 to 186.0 deg-C
Specification Test Results
About this item
:
:408.50 g/mol
CAS Number:550378-78-4
Formula:C₃₀H₂₀N₂
MDL Number:MFCD09836836