Inserisci ordine
Italy
ContactUsLinkComponent
(S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane ≥95.0% (by GC)
Irritant
(S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane ≥95.0% (by GC)
Codice catalogo TCIAD4077-1G
Supplier:  TCI
CAS Number:  
Irritant
(S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane ≥95.0% (by GC)
Codice catalogo TCIAD4077-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Conf:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Specific rotation [a]20/D:
    +4.0 to +6.0 deg (C=15, CHCL3)

Specification Test Results

About this item

 :
 :202.29 g/mol
 :GHS07
CAS Number:151223-12-0
Formula:C₁₁H₂₂O₃