
(R)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane ≥95.0% (by GC)
Catalog # TCIAD4076-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Optical purity (GC):min. 98.0 ee%
- Specific rotation [a]20/D:-4.0 to -6.0 deg (C=15, CHCl3)
Specification Test Results
About this item
:
:202.29 g/mol
:GHS07
CAS Number:1820574-15-9
Formula:C₁₁H₂₂O₃