Order Entry
Italy
ContactUsLinkComponent
Diethyl (1,3-dithian-2-yl)phosphonate ≥93.0% (by GC)
Diethyl (1,3-dithian-2-yl)phosphonate ≥93.0% (by GC)
Catalog # TCIAD4074-5G
Supplier:  TCI
CAS Number:  
Diethyl (1,3-dithian-2-yl)phosphonate ≥93.0% (by GC)
Catalog # TCIAD4074-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to lump
  • Purity (GC):
    min. 93.0 %

Specification Test Results

About this item

 :
 :256.31 g/mol
Boiling Pt:134°C
CAS Number:62999-73-9
Formula:C₈H₁₇O₃PS₂