Order Entry
Italy
ContactUsLinkComponent
 
 
2,5-Dibromohydroquinone ≥98.0% (by GC, titration analysis)
 :  TCI
 
 
2,5-Dibromohydroquinone ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    191.0 to 195.0 deg-C

 

Frequently Bought Together