
5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:174508-31-7
:
Formula:C₆H₄Br₂O₂S
MDL Number:MFCD01651770
:299.9700 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:94.0 to 99.0 deg-C
Specification Test Results
Frequently Bought Together