
2,5-Dichloropyrimidine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:22536-67-0
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00673131
:148.97 g/mol
Boiling Pt:190°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:54.0 to 58.0 deg-C
Specification Test Results
Frequently Bought Together