
1,1-Dimethylpiperidinium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24307-26-4
:
Formula:C₇H₁₆ClN
MDL Number:MFCD00055543
:149.66 g/mol
Merck Index:14,05857
EINECS:246-147-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together