3-(Dimethylamino)-1-(2-thienyl)-1-propanone hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAD4011-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:176.0 to 182.0 deg-C
Specification Test Results
About this item
:
:219.73 g/mol
CAS Number:5424-47-5
Formula:C₉H₁₃NOS
MDL Number:MFCD00126349