
3-(Dimethylamino)-1-(2-thienyl)-1-propanol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13636-02-7
:
Formula:C₉H₁₅NOS
MDL Number:MFCD09701963
:185.29 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:70.0 to 74.0 deg-C
Specification Test Results
Frequently Bought Together